3-Methyl-1-(1-methyl-1H-1,2,4-triazol-5-yl)butan-1-one

Supplier: Matrix Scientific

063926-500MG
101926-646EA 183.08 USD
101926-646
3-Methyl-1-(1-methyl-1H-1,2,4-triazol-5-yl)butan-1-one
3-Methyl-1-(1-methyl-1H-1,2,4-triazol-5-yl)butan-1-one

Hazard: Irritant

Formula: C₈H₁₃N₃O
MDL Number: MFCD09864479
CAS Number: 959239-60-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR