1-(Azidomethyl)-4-methylbenzene

Supplier: Matrix Scientific

063922-500MG
101926-638EA 484.62 USD
101926-638
1-(Azidomethyl)-4-methylbenzene
1-(Azidomethyl)-4-methylbenzene

Hazard: Irritant

Formula: C₈H₉N₃
MDL Number: MFCD11637154
CAS Number: 17271-89-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR