1-(Azidomethyl)-2-methylbenzene

Supplier: Matrix Scientific

063921-500MG
101926-636EA 484.62 USD
101926-636
1-(Azidomethyl)-2-methylbenzene
1-(Azidomethyl)-2-methylbenzene

Hazard: Irritant

Formula: C₈H₉N₃
MDL Number: MFCD11637152
CAS Number: 126799-83-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR