tert-Butyl-3-allyl-3-aminohex-5-enoate

Supplier: Matrix Scientific

063916-500MG
101926-626EA 364.62 USD
101926-626
tert-Butyl-3-allyl-3-aminohex-5-enoate
tert-Butyl-3-allyl-3-aminohex-5-enoate

Hazard: Irritant

Formula: C₁₃H₂₃NO₂
MW: 225.33 g/mol
MDL Number: MFCD16618441
CAS Number: 1228552-15-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR