3-Bromo-5-cyclohexyl-1H-1,2,4-triazole

Supplier: Matrix Scientific

063907-500MG
101926-612EA 364.62 USD
101926-612
3-Bromo-5-cyclohexyl-1H-1,2,4-triazole
3-Bromo-5-cyclohexyl-1H-1,2,4-triazole

Hazard: Irritant

Formula: C₈H₁₂BrN₃
MDL Number: MFCD16556137
CAS Number: 1227465-62-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR