2-[2-(Piperidin-1-ylsulfonyl)ethyl]-1H-isoindole-1,3(2H)-dione

Supplier: Matrix Scientific

063900-500MG
101926-598EA 364.62 USD
101926-598
2-[2-(Piperidin-1-ylsulfonyl)ethyl]-1H-isoindole-1,3(2H)-dione
2-[2-(Piperidin-1-ylsulfonyl)ethyl]-1H-isoindole-1,3(2H)-dione

Hazard: Irritant

Formula: C₁₅H₁₈N₂O₄S
MDL Number: MFCD08339556
CAS Number: 36257-55-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR