2-(1,3-Dioxo-1,3-dihydro-2H-isoindol-2-yl)ethanesulfonamide

Supplier: Matrix Scientific

063899-500MG
101926-596EA 484.62 USD
101926-596
2-(1,3-Dioxo-1,3-dihydro-2H-isoindol-2-yl)ethanesulfonamide
2-(1,3-Dioxo-1,3-dihydro-2H-isoindol-2-yl)ethanesulfonamide

Hazard: Irritant

Formula: C₁₀H₁₀N₂O₄S
MDL Number: MFCD00434121
CAS Number: 4443-23-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR