2-Hydroxyhexahydro-1H-isoindole-1,3(2H)-dione

Supplier: Matrix Scientific

063896-500MG
101926-590EA 484.62 USD
101926-590
2-Hydroxyhexahydro-1H-isoindole-1,3(2H)-dione
2-Hydroxyhexahydro-1H-isoindole-1,3(2H)-dione

Hazard: Irritant

Formula: C₈H₁₁NO₃
MDL Number: MFCD03085911
CAS Number: 18886-85-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR