1-Benzyl-1H-indole-6-carbonitrile

Supplier: Matrix Scientific

063894-500MG
101926-586EA 387.69 USD
101926-586
1-Benzyl-1H-indole-6-carbonitrile
1-Benzyl-1H-indole-6-carbonitrile

Hazard: Irritant

Formula: C₁₆H₁₂N₂
MDL Number: MFCD15200961
CAS Number: 1030423-43-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR