4-Nitrophenyl-N-[(benzyloxy)carbonyl]-L-serinate

Supplier: Matrix Scientific

063890-500MG
101926-580EA 243.08 USD
101926-580
4-Nitrophenyl-N-[(benzyloxy)carbonyl]-L-serinate
4-Nitrophenyl-N-[(benzyloxy)carbonyl]-L-serinate

Hazard: Irritant

Formula: C₁₇H₁₆N₂O₇
MW: 360.33 g/mol
MDL Number: MFCD16618434
CAS Number: 1676-91-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR