Dibenzo[b,d]furan-3-ol

Supplier: Matrix Scientific

063886-500MG
101926-572EA 407.69 USD
101926-572
Dibenzo[b,d]furan-3-ol
Dibenzo[b,d]furan-3-ol

Hazard: Irritant

Formula: C₁₂H₈O₂
MDL Number: MFCD00094159
CAS Number: 20279-16-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR