[(4-Nitrophenyl)thio]acetonitrile

Supplier: Matrix Scientific

063880-500MG
101926-560EA 183.08 USD
101926-560
[(4-Nitrophenyl)thio]acetonitrile
[(4-Nitrophenyl)thio]acetonitrile

Hazard: Irritant

Formula: C₈H₆N₂O₂S
MDL Number: MFCD01210370
CAS Number: 18527-26-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR