(5-Methyl-2-nitrophenyl)methanol

Supplier: Matrix Scientific

063879-500MG
101926-558EA 364.62 USD
101926-558
(5-Methyl-2-nitrophenyl)methanol
(5-Methyl-2-nitrophenyl)methanol

Hazard: Irritant

Formula: C₈H₉NO₃
MDL Number: MFCD00007370
CAS Number: 66424-92-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR