Ethyl-4-amino-1-methyl-1H-pyrazole-3-carboxylate

Supplier: Matrix Scientific

063866-500MG
101926-530EA 364.62 USD
101926-530
Ethyl-4-amino-1-methyl-1H-pyrazole-3-carboxylate
Ethyl-4-amino-1-methyl-1H-pyrazole-3-carboxylate

Hazard: Irritant

Formula: C₇H₁₁N₃O₂
MDL Number: MFCD16618430
CAS Number: 1228552-85-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR