2-Ethylquinazolin-4(3H)-one

Supplier: Matrix Scientific

063863-500MG
101926-524EA 243.08 USD
101926-524
2-Ethylquinazolin-4(3H)-one
2-Ethylquinazolin-4(3H)-one

Hazard: Irritant

Formula: C₁₀H₁₀N₂O
MW: 174.2 g/mol
MDL Number: MFCD00247024
CAS Number: 3137-64-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR