3-Acetyl-2-oxo-2H-chromen-4-yldifluoridoborate

Supplier: Matrix Scientific

063856-500MG
101926-510EA 484.62 USD
101926-510
3-Acetyl-2-oxo-2H-chromen-4-yldifluoridoborate
3-Acetyl-2-oxo-2H-chromen-4-yldifluoridoborate

Hazard: Irritant

Formula: C₁₁H₇BF₂O₄
MDL Number: MFCD01948906
CAS Number: 873326-02-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR