3-Acetyl-6-methyl-2-oxo-2H-pyran-4-yldifluoridoborate

Supplier: Matrix Scientific

063855-500MG
101926-508EA 484.62 USD
101926-508
3-Acetyl-6-methyl-2-oxo-2H-pyran-4-yldifluoridoborate
3-Acetyl-6-methyl-2-oxo-2H-pyran-4-yldifluoridoborate

Hazard: Irritant

Formula: C₈H₇BF₂O₄
MDL Number: MFCD16618428
CAS Number: 67977-10-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR