4-Hydroxy-3-[(2E)-3-(4-methoxyphenyl)prop-2-enoyl]-2H-chromen-2-one

Supplier: Matrix Scientific

063851-500MG
101926-500EA 484.62 USD
101926-500
4-Hydroxy-3-[(2E)-3-(4-methoxyphenyl)prop-2-enoyl]-2H-chromen-2-one
4-Hydroxy-3-[(2E)-3-(4-methoxyphenyl)prop-2-enoyl]-2H-chromen-2-one

Hazard: Irritant

Formula: C₁₉H₁₄O₅
MDL Number: MFCD03019891
CAS Number: 1022282-99-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR