7,7-Dimethyl-2-thioxo-1,5,7,8-tetrahydro-2H-pyrano[4,3-b]pyridine-3-carbonitrile

Supplier: Matrix Scientific

063847-500MG
101926-492EA 484.62 USD
101926-492
7,7-Dimethyl-2-thioxo-1,5,7,8-tetrahydro-2H-pyrano[4,3-b]pyridine-3-carbonitrile
7,7-Dimethyl-2-thioxo-1,5,7,8-tetrahydro-2H-pyrano[4,3-b]pyridine-3-carbonitrile

Hazard: Irritant

Formula: C₁₁H₁₂N₂OS
MDL Number: MFCD01560876
CAS Number: 144291-93-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR