1-(1-Hydroxy-2-pyridin-3-yl-1H-benzimidazol-6-yl)ethanone

Supplier: Matrix Scientific

063837-500MG
101926-476EA 484.62 USD
101926-476
1-(1-Hydroxy-2-pyridin-3-yl-1H-benzimidazol-6-yl)ethanone
1-(1-Hydroxy-2-pyridin-3-yl-1H-benzimidazol-6-yl)ethanone

Hazard: Irritant

Formula: C₁₄H₁₁N₃O₂
MDL Number: MFCD16618422
CAS Number: 1228551-84-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR