Methyl (2Z)-3-amino-3-(benzoylamino)-2-(chloroacetyl)acrylate

Supplier: Matrix Scientific

063835-500MG
101926-472EA 364.62 USD
101926-472
Methyl (2Z)-3-amino-3-(benzoylamino)-2-(chloroacetyl)acrylate
Methyl (2Z)-3-amino-3-(benzoylamino)-2-(chloroacetyl)acrylate

Hazard: Irritant

Formula: C₁₃H₁₃ClN₂O₄
MDL Number: MFCD16618419
CAS Number: 1228572-18-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR