tert-Butyl [2-(5-methyl-1H-indol-3-yl)ethyl]carbamate

Supplier: Matrix Scientific

063832-500MG
101926-468EA 484.62 USD
101926-468
tert-Butyl [2-(5-methyl-1H-indol-3-yl)ethyl]carbamate
tert-Butyl [2-(5-methyl-1H-indol-3-yl)ethyl]carbamate

Hazard: Irritant

Formula: C₁₆H₂₂N₂O₂
MW: 274.37 g/mol
MDL Number: MFCD16618418
CAS Number: 1228552-55-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR