N-[(Z)-Amino(2,4-dioxodihydrofuran-3(2H)-ylidene)methyl]benzamide

Supplier: Matrix Scientific

063831-500MG
101926-466EA 484.62 USD
101926-466
N-[(Z)-Amino(2,4-dioxodihydrofuran-3(2H)-ylidene)methyl]benzamide
N-[(Z)-Amino(2,4-dioxodihydrofuran-3(2H)-ylidene)methyl]benzamide

Hazard: Irritant

Formula: C₁₂H₁₀N₂O₄
MW: 246.22 g/mol
MDL Number: MFCD16618417
CAS Number: 1228572-17-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR