tert-Butyl [2-(5-methoxy-1H-indol-3-yl)ethyl]carbamate

Supplier: Matrix Scientific

063829-500MG
101926-462EA 484.62 USD
101926-462
tert-Butyl [2-(5-methoxy-1H-indol-3-yl)ethyl]carbamate
tert-Butyl [2-(5-methoxy-1H-indol-3-yl)ethyl]carbamate

Hazard: Irritant

Formula: C₁₆H₂₂N₂O₃
MW: 290.37 g/mol
MDL Number: MFCD16618415
CAS Number: 380358-27-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR