Thieno[2,3-d]isothiazol-3(2H)-one-1,1-dioxide

Supplier: Matrix Scientific

063827-500MG
101926-460EA 606.15 USD
101926-460
Thieno[2,3-d]isothiazol-3(2H)-one-1,1-dioxide
Thieno[2,3-d]isothiazol-3(2H)-one-1,1-dioxide

Hazard: Irritant

Formula: C₅H₃NO₃S₂
MW: 189.21 g/mol
MDL Number: MFCD01850954
CAS Number: 59337-94-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR