5-Fluoro-3-methyl-1H-indole-2-carbohydrazide

Supplier: Matrix Scientific

063824-500MG
101926-452EA 364.62 USD
101926-452
5-Fluoro-3-methyl-1H-indole-2-carbohydrazide
5-Fluoro-3-methyl-1H-indole-2-carbohydrazide

Hazard: Irritant

Formula: C₁₀H₁₀FN₃O
MDL Number: MFCD15146560
CAS Number: 1217862-86-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR