2-Methyltetrahydro-2H-pyran-2-carbohydrazide

Supplier: Matrix Scientific

063821-500MG
101926-446EA 530.77 USD
101926-446
2-Methyltetrahydro-2H-pyran-2-carbohydrazide
2-Methyltetrahydro-2H-pyran-2-carbohydrazide

Hazard: Irritant

Formula: C₇H₁₄N₂O₂
MDL Number: MFCD15146553
CAS Number: 1217862-65-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR