4-Hydrazino-2-phenylquinazoline

Supplier: Matrix Scientific

063818-500MG
101926-440EA 606.15 USD
101926-440
4-Hydrazino-2-phenylquinazoline
4-Hydrazino-2-phenylquinazoline

Hazard: Irritant

Formula: C₁₄H₁₂N₄
MDL Number: MFCD07762995
CAS Number: 6484-29-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR