5-Phenyl[1,2,4]triazolo[4,3-c]quinazoline-3-thiol

Supplier: Matrix Scientific

063812-500MG
101926-428EA 484.62 USD
101926-428
5-Phenyl[1,2,4]triazolo[4,3-c]quinazoline-3-thiol
5-Phenyl[1,2,4]triazolo[4,3-c]quinazoline-3-thiol

Hazard: Irritant

Formula: C₁₅H₁₀N₄S
MDL Number: MFCD07763381
CAS Number: 3455-46-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR