4-Hydrazino-2-(3-methoxyphenyl)quinazoline

Supplier: Matrix Scientific

063809-500MG
101926-422EA 606.15 USD
101926-422
4-Hydrazino-2-(3-methoxyphenyl)quinazoline
4-Hydrazino-2-(3-methoxyphenyl)quinazoline

Hazard: Irritant

Formula: C₁₅H₁₄N₄O
MDL Number: MFCD15146547
CAS Number: 1217862-32-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR