6-Amino-1-ethyl-2-thioxo-2,3-dihydropyrimidin-4(1H)-one

Supplier: Matrix Scientific

063808-500MG
101926-420EA 606.15 USD
101926-420
6-Amino-1-ethyl-2-thioxo-2,3-dihydropyrimidin-4(1H)-one
6-Amino-1-ethyl-2-thioxo-2,3-dihydropyrimidin-4(1H)-one

Hazard: Irritant

Formula: C₆H₉N₃OS
MDL Number: MFCD08558475
CAS Number: 15468-21-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR