1-Isobutyl-7-methyl-5-(piperidin-1-ylcarbonyl)pyrido[2,3-d]pyrimidine-2,4(1H,3H)-dione

Supplier: Matrix Scientific

063806-500MG
101926-416EA 364.62 USD
101926-416
1-Isobutyl-7-methyl-5-(piperidin-1-ylcarbonyl)pyrido[2,3-d]pyrimidine-2,4(1H,3H)-dione
1-Isobutyl-7-methyl-5-(piperidin-1-ylcarbonyl)pyrido[2,3-d]pyrimidine-2,4(1H,3H)-dione

Hazard: Irritant

Formula: C₁₈H₂₄N₄O₃
MDL Number: MFCD15146546
CAS Number: 1217863-03-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR