Ethyl-7-cyclopropyl-1-methyl-2,4-dioxo-1,2,3,4-tetrahydropyrido[2,3-d]pyrimidine-5-carboxylate
Supplier: Matrix Scientific
063804-500MG
101926-412EA
484.62
USD
101926-412
Ethyl-7-cyclopropyl-1-methyl-2,4-dioxo-1,2,3,4-tetrahydropyrido[2,3-d]pyrimidine-5-carboxylate
Ethyl-7-cyclopropyl-1-methyl-2,4-dioxo-1,2,3,4-tetrahydropyrido[2,3-d]pyrimidine-5-carboxylate
Hazard: Irritant
Formula:
C₁₄H₁₅N₃O₄ |
MDL Number:
MFCD15146545 CAS Number: 1217863-09-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...