1-(4-Methylphenyl)butane-1,3-dione

Supplier: Matrix Scientific

063801-500MG
101926-406EA 336.92 USD
101926-406
1-(4-Methylphenyl)butane-1,3-dione
1-(4-Methylphenyl)butane-1,3-dione

Hazard: Irritant

Formula: C₁₁H₁₂O₂
MDL Number: MFCD00068483
CAS Number: 4023-79-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR