1-(4-Methoxyphenyl)butane-1,3-dione
Supplier: Matrix Scientific
063800-500MG
101926-404EA
243.08
USD
101926-404
1-(4-Methoxyphenyl)butane-1,3-dione
1-(4-Methoxyphenyl)butane-1,3-dione
Hazard: Irritant
Formula:
C₁₁H₁₂O₃ |
MDL Number:
MFCD00463138 CAS Number: 4023-80-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...