5-(Diethylamino)-2-hydroxybenzaldehyde

Supplier: Matrix Scientific

063793-500MG
101926-390EA 484.62 USD
101926-390
5-(Diethylamino)-2-hydroxybenzaldehyde
5-(Diethylamino)-2-hydroxybenzaldehyde

Hazard: Irritant

Formula: C₁₁H₁₅NO₂
MDL Number: MFCD15146543
CAS Number: 58537-79-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR