2-[(4-Nitrobenzyl)oxy]benzaldehyde

Supplier: Matrix Scientific

063792-500MG
101926-388EA 181.7 USD
101926-388
2-[(4-Nitrobenzyl)oxy]benzaldehyde
2-[(4-Nitrobenzyl)oxy]benzaldehyde

Hazard: Irritant

Formula: C₁₄H₁₁NO₄
MDL Number: MFCD01025493
CAS Number: 17490-72-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR