4-Phenyl-6-(trifluoromethyl)pyrimidin-2-ol

Supplier: Matrix Scientific

063785-500MG
101926-374EA 243.08 USD
101926-374
4-Phenyl-6-(trifluoromethyl)pyrimidin-2-ol
4-Phenyl-6-(trifluoromethyl)pyrimidin-2-ol

Hazard: Irritant

Formula: C₁₁H₇F₃N₂O
MDL Number: MFCD03532398
CAS Number: 67804-93-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR