1-Ethyl-3-methyl-1H-pyrazole-4-carbohydrazide

Supplier: Matrix Scientific

063783-500MG
101926-370EA 243.08 USD
101926-370
1-Ethyl-3-methyl-1H-pyrazole-4-carbohydrazide
1-Ethyl-3-methyl-1H-pyrazole-4-carbohydrazide

Hazard: Irritant

Formula: C₇H₁₂N₄O
MDL Number: MFCD12030797
CAS Number: 1177272-66-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR