5-Amino-4-chloro-1H-pyrazole-3-carboxylic acid

Supplier: Matrix Scientific

063778-500MG
101926-360EA 606.15 USD
101926-360
5-Amino-4-chloro-1H-pyrazole-3-carboxylic acid
5-Amino-4-chloro-1H-pyrazole-3-carboxylic acid

Hazard: Irritant

Formula: C₄H₄ClN₃O₂
MW: 161.55 g/mol
MDL Number: MFCD15146532
CAS Number: 351990-69-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR