1-Ethyl-1H-pyrazole-3-carbonitrile

Supplier: Matrix Scientific

063774-500MG
101926-352EA 335.38 USD
101926-352
1-Ethyl-1H-pyrazole-3-carbonitrile
1-Ethyl-1H-pyrazole-3-carbonitrile

Hazard: Irritant

Formula: C₆H₇N₃
MDL Number: MFCD04969701
CAS Number: 1006471-40-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR