4-Chloro-3,5-dimethylisoxazole

Supplier: Matrix Scientific

063771-500MG
101926-346EA 364.62 USD
101926-346
4-Chloro-3,5-dimethylisoxazole
4-Chloro-3,5-dimethylisoxazole

Hazard: Irritant

Formula: C₅H₆ClNO
MDL Number: MFCD14702892
CAS Number: 10557-86-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR