1-tert-Butyl-4-ethyl-4-propylpiperidine-1,4-dicarboxylate

Supplier: Matrix Scientific

063766-500MG
101926-336EA 364.62 USD
101926-336
1-tert-Butyl-4-ethyl-4-propylpiperidine-1,4-dicarboxylate
1-tert-Butyl-4-ethyl-4-propylpiperidine-1,4-dicarboxylate

Hazard: Irritant

Formula: C₁₆H₂₉NO₄
MDL Number: MFCD15146527
CAS Number: 1033819-09-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR