tert-Butyl (5-bromo-2-hydroxybenzyl)-(2-hydroxyethyl)carbamate
Supplier: Matrix Scientific
063763-500MG
101926-330EA
243.08
USD
101926-330
tert-Butyl (5-bromo-2-hydroxybenzyl)-(2-hydroxyethyl)carbamate
tert-Butyl (5-bromo-2-hydroxybenzyl)-(2-hydroxyethyl)carbamate
Hazard: Irritant
Formula:
C₁₄H₂₀BrNO₄ |
MDL Number:
MFCD15146525 CAS Number: 1217862-54-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...