1-(tert-Butoxycarbonyl)-4-ethylpiperidine-4-carboxylic acid

Supplier: Matrix Scientific

063761-500MG
101926-326EA 160 USD
101926-326
1-(tert-Butoxycarbonyl)-4-ethylpiperidine-4-carboxylic acid
1-(tert-Butoxycarbonyl)-4-ethylpiperidine-4-carboxylic acid

Hazard: Irritant

Formula: C₁₃H₂₃NO₄
MDL Number: MFCD08062529
CAS Number: 188792-67-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR