[3-(Methoxymethoxy)phenyl](pyridin-4-yl)methanone

Supplier: Matrix Scientific

063748-500MG
101926-302EA 243.08 USD
101926-302
[3-(Methoxymethoxy)phenyl](pyridin-4-yl)methanone
[3-(Methoxymethoxy)phenyl](pyridin-4-yl)methanone

Hazard: Irritant

Formula: C₁₄H₁₃NO₃
MDL Number: MFCD09055153
CAS Number: 938458-57-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR