1,3-Benzodioxol-5-yl(phenyl)methanone

Supplier: Matrix Scientific

063747-500MG
101926-300EA 146.15 USD
101926-300
1,3-Benzodioxol-5-yl(phenyl)methanone
1,3-Benzodioxol-5-yl(phenyl)methanone

Hazard: Irritant

Formula: C₁₄H₁₀O₃
MDL Number: MFCD01333932
CAS Number: 54225-86-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR