(4,5-Diphenyl-1H-pyrazol-1-yl)acetic acid

Supplier: Matrix Scientific

063739-500MG
101926-286EA 364.62 USD
101926-286
(4,5-Diphenyl-1H-pyrazol-1-yl)acetic acid
(4,5-Diphenyl-1H-pyrazol-1-yl)acetic acid

Hazard: Irritant

Formula: C₁₇H₁₄N₂O₂
MDL Number: MFCD12031063
CAS Number: 24301-67-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR