[3-(5-Methyl-4H-1,2,4-triazol-3-yl)phenyl]amine

Supplier: Matrix Scientific

063738-500MG
101926-284EA 364.62 USD
101926-284
[3-(5-Methyl-4H-1,2,4-triazol-3-yl)phenyl]amine
[3-(5-Methyl-4H-1,2,4-triazol-3-yl)phenyl]amine

Hazard: Irritant

Formula: C₉H₁₀N₄
MDL Number: MFCD08059833
CAS Number: 915924-22-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR