5-(Aminomethyl)-N-(4-fluorophenyl)-1,3,4-thiadiazole-2-carboxamide
Supplier: Matrix Scientific
063735-500MG
101926-278EA
484.62
USD
101926-278
5-(Aminomethyl)-N-(4-fluorophenyl)-1,3,4-thiadiazole-2-carboxamide
5-(Aminomethyl)-N-(4-fluorophenyl)-1,3,4-thiadiazole-2-carboxamide
Hazard: Irritant
Formula:
C₁₀H₉FN₄OS |
MDL Number:
MFCD15146517 CAS Number: 1217862-69-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...